Learn More
Glycolic acid, 70%
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Supplier: Thermo Scientific Chemicals 411030010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Glycolic acid | |
| 10.0°C | |
| 113.0°C | |
| C2H4O3 | |
| MFCD00004312 | |
| AEMRFAOFKBGASW-UHFFFAOYSA-N | |
| 2-hydroxyacetic acid | |
| 757 | |
| 76.04 | |
| 1.2700 g/mL | |
| Glass bottle | |
| 1.27 |
| 70% in water | |
| 79-14-1,7732-18-5 | |
| Yellow | |
| CH2OHCOOH | |
| glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech | |
| C(C(=O)O)O | |
| 76.051 | |
| CHEBI:17497 | |
| 70% | |
| 1 kg | |
| Solution |
Chemical Identifiers
| 79-14-1 | |
| 76.051 | |
| AEMRFAOFKBGASW-UHFFFAOYSA-N | |
| 757 | |
| 2-hydroxyacetic acid |
| C2H4O3 | |
| MFCD00004312 | |
| glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech | |
| CHEBI:17497 | |
| C(C(=O)O)O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. May damage fertility. May damage the unborn child.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Danger
EINECSNumber : 201-180-5
RTECSNumber : MC5250000
TSCA : TSCA
RUO – Research Use Only