missing translation for 'onlineSavingsMsg'
Learn More

Glycolic acid, 70%, Thermo Scientific™

Numéro de catalogue. AC411030250
Modifier l'affichage
Click to view available options
25kg, Metal drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
AC411030250 25kg, Metal drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC411030250 Fournisseur Thermo Scientific Chemicals Code fournisseur 411030250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Nom ou substance chimique Glycolic acid
Nom Remarque 70% in water
CAS 79-14-1, 7732-18-5
Formule moléculaire C2H4O3
Formule linéaire CH2OHCOOH
Numéro MDL MFCD00004312
Densité 1.2700g/mL
Synonyme glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech
Clé InChI AEMRFAOFKBGASW-UHFFFAOYSA-N
SOURIRES C(C(=O)O)O
Nom de l’IUPAC 2-hydroxyacetic acid
Poids moléculaire (g/mol) 76.051
PubChem CID 757
ChEBI CHEBI:17497
Poids de formule 76.04
Pourcentage de pureté 70%
Emballage Metal drum
Gravité spécifique 1.27
Viscosité 11.28 mPa.s (16°C)
Quantité 25kg
Afficher plus de résultats Afficher moins de résultats
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. May damage fertility. May damage the unborn child.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 201-180-5
RTECSNumber MC5250000
TSCA TSCA

RUO – Research Use Only