missing translation for 'onlineSavingsMsg'
Learn More

Glycolaldehyde dimethyl acetal, 98%, stab. with ca 0.1% sodium carbonate

Catalog No. AAL0709903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0709903 1 g
AAL0709906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0709903 Supplier Thermo Scientific Chemicals Supplier No. L0709903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30934-97-5 | C4H10O3 | 106.12 g/mol

Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 30934-97-5
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00051799
InChI Key NYPNCQTUZYWFGG-UHFFFAOYSA-N
Synonym glycolaldehyde dimethyl acetal, ethanol, 2,2-dimethoxy, unii-u5use33y47, 2,2-dimethoxy-ethanol, 2,2-dimethoxy ethanol, glycolaldehydedimethylacetal, acmc-20aksv, 2,2-dimethoxyethan-1-ol, hydroxyacetaldehyde dimethylacetal, 2-hydroxyacetaldehyde dimethyl acetal
PubChem CID 542381
IUPAC Name 2,2-dimethoxyethanol
SMILES COC(CO)OC

Specifications

Melting Point <-76°C
Density 1.05
Boiling Point 68°C (21 mmHg)
Flash Point 66°C (150°F)
Refractive Index 1.413
Quantity 1 g
Beilstein 1697583
Solubility Information Miscible with water.
Formula Weight 106.12
Percent Purity 98%
Chemical Name or Material Glycolaldehyde dimethyl acetal
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 250-398-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only