missing translation for 'onlineSavingsMsg'
Learn More

Glycolaldehyde diethyl acetal, stab. with ca 0.1% sodium carbonate, 98%

Catalog No. AAL0628218
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0628218 50 g
AAL0628204 2 g
AAL0628209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0628218 Supplier Thermo Scientific Chemicals Supplier No. L0628218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 621-63-6 | C6H14O3 | 134.175 g/mol

Glycolaldehyde diethyl acetal is used in the preparation of neooxazolomycin, which is a part of the oxazolomycin family of antibiotics. It is also used in the preparation of pyrimidine based inhibitors of phosphodiesterase 7.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolaldehyde diethyl acetal is used in the preparation of neooxazolomycin, which is a part of the oxazolomycin family of antibiotics. It is also used in the preparation of pyrimidine based inhibitors of phosphodiesterase 7.

Solubility
Miscible with water, chloroform and ethyl acetate.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acid and bases.

Chemical Identifiers

CAS 621-63-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00051486
InChI Key IKKUKDZKIIIKJK-UHFFFAOYSA-N
Synonym ethanol, 2,2-diethoxy, glycolaldehyde diethyl acetal, 2,2-diethoxy-ethanol, 2,2-diethoxyethan-1-ol, glycolaldehyde, diethyl acetal, 2,2-diethoxy alcohol, glycolaldehyde diethyl acetal, stab. with ca sodium carbonate, ethanol,2-diethoxy, acmc-20a2bj
PubChem CID 12129
IUPAC Name 2,2-diethoxyethanol
SMILES CCOC(CO)OCC

Specifications

Boiling Point 166°C to 167°C
Flash Point 62°C (143°F)
Refractive Index 1.416
Quantity 50 g
Beilstein 773727
Solubility Information Miscible with water,chloroform and ethyl acetate.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material Glycolaldehyde diethyl acetal, stabilized with ≈0.1% sodium carbonate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 210-697-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only