missing translation for 'onlineSavingsMsg'
Learn More

Glycol sulfite, 98%

Catalog No. AAH6013314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6013314 25 g
AAH6013306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6013314 Supplier Thermo Scientific Chemicals Supplier No. H6013314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3741-38-6 | C2H4O3S | 108.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3741-38-6
Molecular Formula C2H4O3S
Molecular Weight (g/mol) 108.11
MDL Number MFCD00005354
InChI Key WDXYVJKNSMILOQ-UHFFFAOYSA-N
Synonym ethylene sulfite, glycol sulfite, 1,3,2-dioxathiolane, 2-oxide, glycolsulfite, 1,2-ethylene sulfite, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulphite, sulfurous acid, cyclic ester with ethylene glycol
PubChem CID 77342
IUPAC Name 1,3,2-dioxathiolane 2-oxide
SMILES O=S1OCCO1

Specifications

Density 1.426
Boiling Point 159°C
Flash Point 79°C (174°F)
Refractive Index 1.445
Quantity 25 g
Beilstein 1237109
Formula Weight 108.12
Percent Purity 98%
Chemical Name or Material Glycol sulfite
Hazard Category H227-H301-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P310-P305+P351+P338-P330-P370+P378q-P501c
EINECSNumber 223-131-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only