MW 0 g/mol. Branched polymer of glucose synthesized by animal cells for energy storage and release. It is an analog of starch, has a similar structure to amylopectin, and functions as a secondary long-term energy storage. Used as a carrier substance for nucleic acid precipitation. Acts as an inert carrier which significantly increases the recovery of extracted DNA and RNA by ethanol precipitation.
The product is subject to the following: Abcam Restricted Use Statement