missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine Zinc Salt Monohydrate 97.0+%, TCI America™
$175.22 - $385.86
Chemical Identifiers
| CAS | 14281-83-5 |
|---|---|
| Molecular Formula | C4H8N2O4Zn |
| Molecular Weight (g/mol) | 213.498 |
| MDL Number | MFCD00070561 |
| InChI Key | UOXSXMSTSYWNMH-UHFFFAOYSA-L |
| Synonym | Zinc Glycinate |
| PubChem CID | 151910 |
| IUPAC Name | zinc;2-aminoacetate |
| SMILES | C(C(=O)[O-])N.C(C(=O)[O-])N.[Zn+2] |
Chemical Identifiers
| 14281-83-5 | |
| 213.498 | |
| UOXSXMSTSYWNMH-UHFFFAOYSA-L | |
| 151910 | |
| C(C(=O)[O-])N.C(C(=O)[O-])N.[Zn+2] |
| C4H8N2O4Zn | |
| MFCD00070561 | |
| Zinc Glycinate | |
| zinc;2-aminoacetate |
Specifications
| 14281-83-5 | |
| C4H8N2O4Zn | |
| 5 g | |
| UOXSXMSTSYWNMH-UHFFFAOYSA-L | |
| zinc;2-aminoacetate | |
| 151910 | |
| ≥97.0% (T) | |
| Glycine Zinc Salt Monohydrate [for Protein Research] |
| White | |
| MFCD00070561 | |
| Zinc Glycinate | |
| C(C(=O)[O-])N.C(C(=O)[O-])N.[Zn+2] | |
| 213.498 | |
| 213.50 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1201
TSCA : No