missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine Zinc Salt Monohydrate 97.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 213.50 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Glycine Zinc Salt Monohydrate [for Protein Research] |
Safety and Handling
| EINECSNumber | (2)-1201 |
| TSCA | No |