missing translation for 'onlineSavingsMsg'
Learn More

Glycine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7130212
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02541G 1 g
G02545G 5 g
G025425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G02541G Supplier TCI America Supplier No. G02541G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27532-96-3
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD00058255
InChI Key OSWULUXZFOQIRU-UHFFFAOYSA-N
Synonym h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride
PubChem CID 2725036
IUPAC Name tert-butyl 2-aminoacetate hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CN

Specifications

Color White
Quantity 1 g
Formula Weight 167.63
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Glycine tert-Butyl Ester Hydrochloride