missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine tert-butyl ester hydrochloride, 99%
CAS: 27532-96-3 | C6H14NO2 | 132.18 g/mol
$80.25 - $295.96
Chemical Identifiers
| CAS | 27532-96-3 |
|---|---|
| Molecular Formula | C6H14ClNO2 |
| Molecular Weight (g/mol) | 167.63 |
| MDL Number | MFCD00058255 |
| InChI Key | OSWULUXZFOQIRU-UHFFFAOYSA-N |
| Synonym | h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride |
| PubChem CID | 2725036 |
| IUPAC Name | tert-butyl 2-aminoacetate hydrochloride |
| SMILES | Cl.CC(C)(C)OC(=O)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1711603
|
Thermo Scientific Chemicals
L1711603 |
1 g |
Each for $80.25
|
|
|||||
|
AAL1711606
|
Thermo Scientific Chemicals
L1711606 |
5 g |
Each for $295.96
|
|
|||||
Description
Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.
Solubility
Soluble in methanol and water
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 27532-96-3 | |
| 167.63 | |
| OSWULUXZFOQIRU-UHFFFAOYSA-N | |
| 2725036 | |
| Cl.CC(C)(C)OC(=O)CN |
| C6H14ClNO2 | |
| MFCD00058255 | |
| h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride | |
| tert-butyl 2-aminoacetate hydrochloride |
Specifications
| ∽140°C (decomposition) | |
| C6H14ClNO2 | |
| 3989048 | |
| Soluble in methanol and water | |
| Cl.CC(C)(C)OC(=O)CN | |
| 1 g | |
| 2725036 | |
| Hygroscopic | |
| Glycine tert-butyl ester hydrochloride |
| 27532-96-3 | |
| MFCD00058255 | |
| h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride | |
| OSWULUXZFOQIRU-UHFFFAOYSA-N | |
| tert-butyl 2-aminoacetate hydrochloride | |
| 167.63 | |
| 167.64 | |
| 99% |
Safety and Handling
EINECSNumber : 214-223-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only