missing translation for 'onlineSavingsMsg'
Learn More

Glycine tert-butyl ester hydrochloride, 99%

Catalog No. AAL1711603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1711603 1 g
AAL1711606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1711603 Supplier Thermo Scientific Chemicals Supplier No. L1711603

CAS: 27532-96-3 | C6H14NO2 | 132.18 g/mol

Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

Solubility
Soluble in methanol and water

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 27532-96-3
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD00058255
InChI Key OSWULUXZFOQIRU-UHFFFAOYSA-N
Synonym h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride
PubChem CID 2725036
IUPAC Name tert-butyl 2-aminoacetate hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CN

Specifications

Melting Point ∽140°C (decomposition)
Beilstein 3989048
Solubility Information Soluble in methanol and water
Quantity 1 g
Formula Weight 167.64
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material Glycine tert-butyl ester hydrochloride
EINECSNumber 214-223-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only