missing translation for 'onlineSavingsMsg'
Learn More

Promega Glycine, Molecular Biology Grade, Promega™

Catalog No. PRH5073
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRH5073 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRH5073 Supplier Promega Supplier No. H5073

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An amino acid used in the preparation of some electrophoresis buffers.

  • Quality tested and certified free of DNase, RNase and protease activity

Specifications

CAS 56-40-6
Color White
Physical Form Crystalline
Quantity 1 kg
Molecular Weight (g/mol) 75.067