missing translation for 'onlineSavingsMsg'
Learn More

Glycine hydrochloride, 98%

Catalog No. AAA1216236
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1216236 500 g
AAA1216222 100 g
AAA121620E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1216236 Supplier Thermo Scientific Chemicals Supplier No. A1216236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6000-43-7 | C2H6ClNO2 | 111.53 g/mol

Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine hydrochloride is an inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Solubility
Soluble in water (>100 g/L).

Notes
Hygroscopic. Store away from strong oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 6000-43-7
Molecular Formula C2H6ClNO2
Molecular Weight (g/mol) 111.53
MDL Number MFCD00012872
InChI Key IVLXQGJVBGMLRR-UHFFFAOYSA-N
Synonym glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride
PubChem CID 22316
IUPAC Name 2-aminoacetic acid;hydrochloride
SMILES [H+].[Cl-].NCC(O)=O

Specifications

Melting Point 175°C to 178°C
Beilstein 3909617
Merck Index 14,4491
Solubility Information Soluble in water (>100g/L).
Quantity 500 g
Odor Odorless
Formula Weight 111.53
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material Glycine hydrochloride
EINECSNumber 227-841-8
RTECSNumber MC0560000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only