missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine hydrochloride, 98%

Catalog No. p-139012
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411011000 100 g Glass Bottle
AC411015000 500 g Glass Bottle
AC411010025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC411011000 Supplier Thermo Scientific Chemicals Supplier No. 411011000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6000-43-7 | C2H6ClNO2 | 111.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6000-43-7
Molecular Formula C2H6ClNO2
Molecular Weight (g/mol) 111.53
MDL Number MFCD00012872
InChI Key IVLXQGJVBGMLRR-UHFFFAOYSA-N
Synonym glycine hydrochloride, 2-aminoacetic acid hydrochloride, glycine, hydrochloride, glycocoll hydrochloride, aminoacetic acid hydrochloride, glycine-hcl, unii-225zlc74hx, ccris 3355, glycine hcl, glycine, chloride
PubChem CID 22316
IUPAC Name 2-aminoacetic acid;hydrochloride
SMILES [H+].[Cl-].NCC(O)=O

Specifications

Melting Point 178.0°C to 182.0°C
pH 1 to 2 (10% aq. soln.)
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula NH2CH2COOH·HCl
Beilstein 04, 340
Merck Index 15, 4526
Solubility Information Solubility in water: soluble.
Water 1% Max. (K.F.)
Quantity 100 g
Formula Weight 111.53
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Glycine hydrochloride, 99%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 227-841-8
RTECSNumber MC0560000
TSCA TSCA

RUO – Research Use Only