missing translation for 'onlineSavingsMsg'
Learn More

Glycine, For Chlorine, Chlorine Dioxide, 20% (w/v), LabChem™

Catalog No. LC148851
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC148851 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC148851 Supplier LabChem Supplier No. LC148851
Only null left

Specifications

Color Clear, colorless to slight yellow
Format Liquid
Packaging Poly Bottle
Quantity 500 mL
Storage Requirements Refrigerate
Type Glycine