missing translation for 'onlineSavingsMsg'
Learn More

Glycine Ethyl Ester Hydrochloride 99.0+%, TCI America™

Catalog No. G010225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G010225G 25 g
G0102500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G010225G Supplier TCI America Supplier No. G010225G

Chemical Identifiers

CAS 623-33-6
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.579
MDL Number MFCD00012871
InChI Key TXTWXQXDMWILOF-UHFFFAOYSA-N
Synonym glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor
PubChem CID 2723640
IUPAC Name ethyl 2-aminoacetate;hydrochloride
SMILES CCOC(=O)CN.Cl

Specifications

Melting Point 146°C
Color White
Quantity 25 g
Formula Weight 139.58
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycine Ethyl Ester Hydrochloride
EINECSNumber (2)-1200
RTECSNumber MC0525000
TSCA Yes