missing translation for 'onlineSavingsMsg'
Learn More

Glycine ethyl ester hydrochloride, 99%

CAS: 623-33-6 | C4H10ClNO2 | 139.579 g/mol

Supplier:  Thermo Scientific Chemicals A103150E

Quantity 2500 g

 View more versions of this product

Catalog No. AAA103150E


Only null left

Description

Description

Glycine ethyl ester (GEE) is used in conjunction with N-(3-Dimethylaminopropyl)-N?-ethylcarbodiimide (EDC) for carboxyl-foot printing studies of proteins. The GEE/EDC protocol effects specific derivatization of glutamate and aspartate carboxyl side chains on intact proteins. This reaction is readily done under aqueous conditions at physiological pH. It is mainly used as a pharmaceutical raw material. Glycine ethyl ester hydrochloride is an important intermediate for preparing chrysanthemic acid or two chlorine chrysanthemic acid and is mainly used in the synthesis of anti-inflammatory drugs, but also can be used for the synthesis of four imidazole acetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine ethyl ester (GEE) is used in conjunction with N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide (EDC) for carboxyl-foot printing studies of proteins. The GEE/EDC protocol effects specific derivatization of glutamate and aspartate carboxyl side chains on intact proteins. This reaction is readily done under aqueous conditions at physiological pH. It is mainly used as a pharmaceutical raw material. Glycine ethyl ester hydrochloride is an important intermediate for preparing chrysanthemic acid or two chlorine chrysanthemic acid and is mainly used in the synthesis of anti-inflammatory drugs, but also can be used for the synthesis of four imidazole acetic acid.

Solubility
Soluble in water (>1000 mg/ml at 20°C), ethanol, DMSO, and methanol.

Notes
Hygroscopic. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Specifications

Chemical Identifiers

623-33-6
139.579
TXTWXQXDMWILOF-UHFFFAOYSA-N
2723640
CCOC(=O)CN.Cl
C4H10ClNO2
MFCD00012871
glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor
ethyl 2-aminoacetate;hydrochloride

Specifications

144°C to 148°C
623-33-6
MFCD00012871
glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor
TXTWXQXDMWILOF-UHFFFAOYSA-N
ethyl 2-aminoacetate;hydrochloride
139.579
139.58
99%
Glycine ethyl ester hydrochloride
C4H10ClNO2
3594034
Soluble in water (>1000mg/ml at 20°C),ethanol,DMSO,and methanol.
CCOC(=O)CN.Cl
2500 g
2723640
Hygroscopic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

EINECSNumber : 210-787-4

RTECSNumber : MC0525000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions