missing translation for 'onlineSavingsMsg'
Learn More

Glycine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™

$29.83 - $67.11

Chemical Identifiers

CAS 1738-76-7
Molecular Formula C16H19NO5S
Molecular Weight (g/mol) 337.39
MDL Number MFCD00035425
InChI Key WJKJXKRHMUXQSL-UHFFFAOYSA-N
Synonym h-gly-obzl.tosoh, glycine benzyl ester p-toluenesulfonate, glycine benzyl ester p-toluenesulfonate salt, benzyl glycinate p-toluenesulfonate, o-benzylglycine toluene-p-sulphonate, h-gly-obzl p-tosylate, gly-obzl.tsoh, aminoacetic acid benzyl ester toluene-4-sulfonic acid salt, glycine benzyl ester p-tosylate, glycine benzyl ester 4-toluenesulfonate
PubChem CID 6451311
IUPAC Name benzyl 2-aminoacetate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B02665G
View Documents
TCI America
B02665G
5 g
Each for $29.83
Only null left
 
B026625G
View Documents
TCI America
B026625G
25 g
Each for $67.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1738-76-7
337.39
WJKJXKRHMUXQSL-UHFFFAOYSA-N
6451311
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CN
C16H19NO5S
MFCD00035425
h-gly-obzl.tosoh, glycine benzyl ester p-toluenesulfonate, glycine benzyl ester p-toluenesulfonate salt, benzyl glycinate p-toluenesulfonate, o-benzylglycine toluene-p-sulphonate, h-gly-obzl p-tosylate, gly-obzl.tsoh, aminoacetic acid benzyl ester toluene-4-sulfonic acid salt, glycine benzyl ester p-tosylate, glycine benzyl ester 4-toluenesulfonate
benzyl 2-aminoacetate;4-methylbenzenesulfonic acid

Specifications

1738-76-7
White
MFCD00035425
h-gly-obzl.tosoh, glycine benzyl ester p-toluenesulfonate, glycine benzyl ester p-toluenesulfonate salt, benzyl glycinate p-toluenesulfonate, o-benzylglycine toluene-p-sulphonate, h-gly-obzl p-tosylate, gly-obzl.tsoh, aminoacetic acid benzyl ester toluene-4-sulfonic acid salt, glycine benzyl ester p-tosylate, glycine benzyl ester 4-toluenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CN
337.39
337.39
Crystalline Powder
135°C
C16H19NO5S
5 g
WJKJXKRHMUXQSL-UHFFFAOYSA-N
benzyl 2-aminoacetate;4-methylbenzenesulfonic acid
6451311
≥98.0% (HPLC,T)
Glycine Benzyl Ester p-Toluenesulfonate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'