missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine benzyl ester p-toluenesulfonate salt, 99%
CAS: 1738-76-7 | C16H19NO5S | 337.39 g/mol
$72.01 - $238.32
Chemical Identifiers
| CAS | 1738-76-7 |
|---|---|
| Molecular Formula | C16H19NO5S |
| Molecular Weight (g/mol) | 337.39 |
| MDL Number | MFCD00035425 |
| InChI Key | WJKJXKRHMUXQSL-UHFFFAOYSA-N |
| Synonym | h-gly-obzl.tosoh, glycine benzyl ester p-toluenesulfonate, glycine benzyl ester p-toluenesulfonate salt, benzyl glycinate p-toluenesulfonate, o-benzylglycine toluene-p-sulphonate, h-gly-obzl p-tosylate, gly-obzl.tsoh, aminoacetic acid benzyl ester toluene-4-sulfonic acid salt, glycine benzyl ester p-tosylate, glycine benzyl ester 4-toluenesulfonate |
| PubChem CID | 6451311 |
| IUPAC Name | benzyl 2-aminoacetate;4-methylbenzenesulfonic acid |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5601606
|
Thermo Scientific Chemicals
H5601606 |
5 g |
Each for $72.01
|
|
|||||
|
AAH5601614
|
Thermo Scientific Chemicals
H5601614 |
25 g |
Each for $238.32
|
|
|||||
Description
Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.
Solubility
Soluble in methanol. (0.1 g/mL)
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Chemical Identifiers
| 1738-76-7 | |
| 337.39 | |
| WJKJXKRHMUXQSL-UHFFFAOYSA-N | |
| 6451311 | |
| CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CN |
| C16H19NO5S | |
| MFCD00035425 | |
| h-gly-obzl.tosoh, glycine benzyl ester p-toluenesulfonate, glycine benzyl ester p-toluenesulfonate salt, benzyl glycinate p-toluenesulfonate, o-benzylglycine toluene-p-sulphonate, h-gly-obzl p-tosylate, gly-obzl.tsoh, aminoacetic acid benzyl ester toluene-4-sulfonic acid salt, glycine benzyl ester p-tosylate, glycine benzyl ester 4-toluenesulfonate | |
| benzyl 2-aminoacetate;4-methylbenzenesulfonic acid |
Specifications
| 1738-76-7 | |
| C16H19NO5S | |
| MFCD00035425 | |
| 3600116 | |
| Soluble in methanol. (0.1g/mL) | |
| CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CN | |
| 337.39 | |
| 337.39 | |
| Glycine benzyl ester p-toluenesulfonate salt |
| 132°C to 134°C | |
| NH2CH2COOCH2C6H5·CH3C6H4SO3H | |
| 5 g | |
| h-gly-obzl.tosoh, glycine benzyl ester p-toluenesulfonate, glycine benzyl ester p-toluenesulfonate salt, benzyl glycinate p-toluenesulfonate, o-benzylglycine toluene-p-sulphonate, h-gly-obzl p-tosylate, gly-obzl.tsoh, aminoacetic acid benzyl ester toluene-4-sulfonic acid salt, glycine benzyl ester p-tosylate, glycine benzyl ester 4-toluenesulfonate | |
| WJKJXKRHMUXQSL-UHFFFAOYSA-N | |
| benzyl 2-aminoacetate;4-methylbenzenesulfonic acid | |
| 6451311 | |
| 99% |
Safety and Handling
EINECSNumber : 217-094-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only