missing translation for 'onlineSavingsMsg'
Learn More

Glycine benzyl ester p-toluenesulfonate salt, 99%

Catalog No. AAH5601606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5601606 5 g
AAH5601614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5601606 Supplier Thermo Scientific Chemicals Supplier No. H5601606
Only null left

CAS: 1738-76-7 | C16H19NO5S | 337.39 g/mol

Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.

Solubility
Soluble in methanol. (0.1 g/mL)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 1738-76-7
Molecular Formula C16H19NO5S
Molecular Weight (g/mol) 337.39
MDL Number MFCD00035425
InChI Key WJKJXKRHMUXQSL-UHFFFAOYSA-N
Synonym h-gly-obzl.tosoh, glycine benzyl ester p-toluenesulfonate, glycine benzyl ester p-toluenesulfonate salt, benzyl glycinate p-toluenesulfonate, o-benzylglycine toluene-p-sulphonate, h-gly-obzl p-tosylate, gly-obzl.tsoh, aminoacetic acid benzyl ester toluene-4-sulfonic acid salt, glycine benzyl ester p-tosylate, glycine benzyl ester 4-toluenesulfonate
PubChem CID 6451311
IUPAC Name benzyl 2-aminoacetate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)COC(=O)CN

Specifications

Melting Point 132°C to 134°C
Linear Formula NH2CH2COOCH2C6H5·CH3C6H4SO3H
Quantity 5 g
Beilstein 3600116
Solubility Information Soluble in methanol. (0.1g/mL)
Formula Weight 337.39
Percent Purity 99%
Chemical Name or Material Glycine benzyl ester p-toluenesulfonate salt
EINECSNumber 217-094-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only