missing translation for 'onlineSavingsMsg'
Learn More

Glycine Anhydride 98.0+%, TCI America™

Catalog No. G0100500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0100500G 500 g
G010025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0100500G Supplier TCI America Supplier No. G0100500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-57-0
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.104
MDL Number MFCD00006009
InChI Key BXRNXXXXHLBUKK-UHFFFAOYSA-N
Synonym 2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
PubChem CID 7817
ChEBI CHEBI:16535
IUPAC Name piperazine-2,5-dione
SMILES C1C(=O)NCC(=O)N1

Specifications

Color White
Quantity 500 g
Formula Weight 114.10
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Glycine Anhydride
RTECSNumber TL6325250
TSCA Yes