missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine anhydride, 98%

CAS: 106-57-0 | C4H6N2O2 | 114.104 g/mol

$107.95 - $323.82

Chemical Identifiers

CAS 106-57-0
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.104
MDL Number MFCD00006009
InChI Key BXRNXXXXHLBUKK-UHFFFAOYSA-N
Synonym 2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
PubChem CID 7817
ChEBI CHEBI:16535
IUPAC Name piperazine-2,5-dione
SMILES C1C(=O)NCC(=O)N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1882214
View Documents Promotion Details
Thermo Scientific Chemicals
A1882214
25 g
Each for $107.95
Only null left
 
AAA1882222
View Documents Promotion Details
Thermo Scientific Chemicals
A1882222
100 g
Each for $323.82
Only null left
 
Description

Description

It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.

Solubility
Soluble in water (142mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

106-57-0
114.104
BXRNXXXXHLBUKK-UHFFFAOYSA-N
7817
piperazine-2,5-dione
C4H6N2O2
MFCD00006009
2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
CHEBI:16535
C1C(=O)NCC(=O)N1

Specifications

∼300°C (decomposition)
C4H6N2O2
112112
2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
BXRNXXXXHLBUKK-UHFFFAOYSA-N
piperazine-2,5-dione
7817
114.1
Glycine anhydride
106-57-0
MFCD00006009
14,7466
Soluble in water (142mg/L).
C1C(=O)NCC(=O)N1
114.104
CHEBI:16535
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-411-5

RTECSNumber : TL6325250

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only