missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine anhydride, 98%

Catalog No. AAA1882214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1882214 25 g
AAA1882222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1882214 Supplier Thermo Scientific Chemicals Supplier No. A1882214

CAS: 106-57-0 | C4H6N2O2 | 114.104 g/mol

It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as an intermediate in the preparation of various pharmaceutical and biologically active compounds. Also used in the synthesis of dipeptide isosteres by cross-metathesis.

Solubility
Soluble in water (142mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 106-57-0
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.104
MDL Number MFCD00006009
InChI Key BXRNXXXXHLBUKK-UHFFFAOYSA-N
Synonym 2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
PubChem CID 7817
ChEBI CHEBI:16535
IUPAC Name piperazine-2,5-dione
SMILES C1C(=O)NCC(=O)N1

Specifications

Melting Point ∼300°C (decomposition)
Beilstein 112112
Merck Index 14,7466
Solubility Information Soluble in water (142mg/L).
Quantity 25 g
Formula Weight 114.1
Percent Purity 99%
Chemical Name or Material Glycine anhydride
EINECSNumber 203-411-5
RTECSNumber TL6325250
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only