missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycidyl Phenyl Ether 99.0+%, TCI America™
$36.57 - $88.56
Chemical Identifiers
| CAS | 122-60-1 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.18 |
| MDL Number | MFCD00005133,MFCD31699959 |
| InChI Key | FQYUMYWMJTYZTK-UHFFFAOYNA-N |
| Synonym | glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl |
| PubChem CID | 31217 |
| ChEBI | CHEBI:82367 |
| IUPAC Name | 2-(phenoxymethyl)oxirane |
| SMILES | C(OC1=CC=CC=C1)C1CO1 |
Chemical Identifiers
| 122-60-1 | |
| 150.18 | |
| FQYUMYWMJTYZTK-UHFFFAOYNA-N | |
| 31217 | |
| 2-(phenoxymethyl)oxirane |
| C9H10O2 | |
| MFCD00005133,MFCD31699959 | |
| glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl | |
| CHEBI:82367 | |
| C(OC1=CC=CC=C1)C1CO1 |
Specifications
| 122-60-1 | |
| 245°C | |
| MFCD00005133,MFCD31699959 | |
| glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl | |
| C(OC1=CC=CC=C1)C1CO1 | |
| 150.18 | |
| CHEBI:82367 | |
| ≥99.0% (GC) | |
| Glycidyl Phenyl Ether |
| Yellow | |
| C9H10O2 | |
| 25 g | |
| FQYUMYWMJTYZTK-UHFFFAOYNA-N | |
| 2-(phenoxymethyl)oxirane | |
| 31217 | |
| 150.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0559&(3)-0594
RTECSNumber : TZ3675000
TSCA : Yes