missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Phenyl Ether 99.0+%, TCI America™

Catalog No. G009825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G009825G 25 g
G0098500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G009825G Supplier TCI America Supplier No. G009825G

Chemical Identifiers

CAS 122-60-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00005133,MFCD31699959
InChI Key FQYUMYWMJTYZTK-UHFFFAOYNA-N
Synonym glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl
PubChem CID 31217
ChEBI CHEBI:82367
IUPAC Name 2-(phenoxymethyl)oxirane
SMILES C(OC1=CC=CC=C1)C1CO1

Specifications

Color Yellow
Boiling Point 245°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycidyl Phenyl Ether
EINECSNumber (3)-0559&(3)-0594
RTECSNumber TZ3675000
TSCA Yes