missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl phenyl ether, 99%

Catalog No. p-7026002
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2176818 50 g
AAB2176830 250 g
AAB217680B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2176818 Supplier Thermo Scientific Chemicals Supplier No. B2176818
Only null left

CAS: 122-60-1 | C9H10O2 | 150.18 g/mol

2-(Phenoxymethyl)oxirane is used in the synthesis of hydrazide-containing inhibitors of HIV-1 integrase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Phenoxymethyl)oxirane is used in the synthesis of hydrazide-containing inhibitors of HIV-1 integrase.

Solubility
Soluble in water (2.4g/L).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 122-60-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00005133,MFCD31699959
InChI Key FQYUMYWMJTYZTK-UHFFFAOYNA-N
Synonym glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl
PubChem CID 31217
ChEBI CHEBI:82367
IUPAC Name 2-(phenoxymethyl)oxirane
SMILES C(OC1=CC=CC=C1)C1CO1

Specifications

Melting Point 3.5°C
Density 1.11
Boiling Point 244°C to 246°C
Flash Point 114°C (237°F)
Odor Sweet
Refractive Index 1.531
Quantity 50 g
Beilstein 2744
Solubility Information Soluble in water (2.4g/L).
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Glycidyl phenyl ether
Show More Show Less
Hazard Category H315-H317-H319-H332-H335-H341-H350
Hazard Statement GHS H Statement
H350-H341-H332-H315-H317-H335
May cause cancer.
Suspected of causing genetic defects.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
EINECSNumber 204-557-2
RTECSNumber TZ3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only