missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Methacrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. M059025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M059025G 25 g
M0590500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M059025G Supplier TCI America Supplier No. M059025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-91-2
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00005137
InChI Key VOZRXNHHFUQHIL-UHFFFAOYSA-N
Synonym glycidyl methacrylate, 2,3-epoxypropyl methacrylate, sy-monomer g, glycidol methacrylate, acriester g, blemmer g, blemmer gma, light ester g, glycidylmethacrylate, 2-methacryloxy methyl oxirane
PubChem CID 7837
IUPAC Name oxiran-2-ylmethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1CO1

Specifications

Color Yellow
Boiling Point 189°C
Quantity 25 g
UN Number 2922
Formula Weight 142.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycidyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1041
RTECSNumber OZ4375000
TSCA Yes
Recommended Storage Refrigerator