missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol

Catalog No. p-7045555
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1113318 50 g
AAL1113330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1113318 Supplier Thermo Scientific Chemicals Supplier No. L1113318
Only null left

CAS: 106-91-2 | C7H10O3 | 142.154 g/mol

Glycidyl methacrylate is commonly used as a monomer for the preparation of epoxy resins. It is also used to provide epoxy functionalization to polyolefins and other acrylate resins. Further, it is also useful as an epoxy resin additive for paint coating formulations and adhesive applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycidyl methacrylate is commonly used as a monomer for the preparation of epoxy resins. It is also used to provide epoxy functionalization to polyolefins and other acrylate resins. Further, it is also useful as an epoxy resin additive for paint coating formulations and adhesive applications.

Solubility
Miscible with benzene, alcohol and ether. Slightly miscible with water.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases and peroxides.

Chemical Identifiers

CAS 106-91-2
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.154
MDL Number MFCD00005137
InChI Key VOZRXNHHFUQHIL-UHFFFAOYSA-N
Synonym glycidyl methacrylate, 2,3-epoxypropyl methacrylate, sy-monomer g, glycidol methacrylate, acriester g, blemmer g, blemmer gma, light ester g, glycidylmethacrylate, 2-methacryloxy methyl oxirane
PubChem CID 7837
IUPAC Name oxiran-2-ylmethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1CO1

Specifications

Melting Point -82°C
Density 1.078
Boiling Point 189°C
Flash Point 83°C (181°F)
Refractive Index 1.45
Quantity 50 g
UN Number UN2810
Beilstein 2506
Solubility Information Miscible with benzene,alcohol and ether. Slightly miscible with water.
Formula Weight 142.16
Percent Purity 97%
Chemical Name or Material Glycidyl methacrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less
Hazard Category H227-H302-H311-H314-H317-H335-H341-H350-H360F-H372
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H317-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 203-441-9
RTECSNumber OZ4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only