missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Glycidyl methacrylate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$2209.75 - $2209.75
Specifications
| CAS | 106-91-2 |
|---|---|
| Density | 1.042 g/mL (at 25°C) |
| Boiling Point | 189°C |
| Molecular Formula | C7H10O3 |
| Linear Formula | C7H10O3 |
Description
Glycidyl methacrylate (GMA) is a polyfunctional monomer. It acts as an adhesion promoting crosslinking co-monomer for acrylic and vinyl resins. It is also a reactive colorless diluent. GMA is soluble in ethanol, acetone, diethyl ether, benzene.Glycidyl methacrylate dextran (GMA) has been reported to be used as a biocompatible hydrogel. In situ polymerization of GMA with trimethylolpropane trimethacrylate to form macroporous sorbents has also been reported. GMA may also be grafted onto polypropylene.Specifications
| 106-91-2 | |
| 189°C | |
| C7H10O3 | |
| MFCD00005137 | |
| 142.15 | |
| Respiratory Tract |
| 1.042 g/mL (at 25°C) | |
| C7H10O3 | |
| n20/D 1.449 (literature) | |
| 2,3-Epoxypropyl methacrylate; Methacrylic acid 2,3-epoxypropyl ester | |
| 97% |