missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Glycidyl methacrylate

Numéro de catalogue. 1512385KG
Encompass
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
1512385KG 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 1512385KG Fournisseur Sigma Aldrich Code fournisseur 1512385KG
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycidyl methacrylate (GMA) is a polyfunctional monomer. It acts as an adhesion promoting crosslinking co-monomer for acrylic and vinyl resins. It is also a reactive colorless diluent. GMA is soluble in ethanol, acetone, diethyl ether, benzene.Glycidyl methacrylate dextran (GMA) has been reported to be used as a biocompatible hydrogel. In situ polymerization of GMA with trimethylolpropane trimethacrylate to form macroporous sorbents has also been reported. GMA may also be grafted onto polypropylene.

Spécifications

CAS 106-91-2
Density 1.042 g/mL (at 25°C)
Boiling Point 189°C
Molecular Formula C7H10O3
Refractive Index n20/D 1.449 (literature)
Linear Formula C7H10O3
Quantity 5 kg
MDL Number MFCD00005137
Synonym 2,3-Epoxypropyl methacrylate; Methacrylic acid 2,3-epoxypropyl ester
Molecular Weight (g/mol) 142.15
Percent Purity 97%
Biological Activity Respiratory Tract
Afficher plus de résultats Afficher moins de résultats