missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Isopropyl Ether 96.0+%, TCI America™

$106.25 - $296.38

Chemical Identifiers

CAS 4016-14-2
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00005139
InChI Key NWLUZGJDEZBBRH-UHFFFAOYSA-N
Synonym glycidyl isopropyl ether, isopropyl glycidyl ether, 2,3-epoxypropyl isopropyl ether, isopropoxymethyl oxirane, 2-isopropoxymethyl oxirane, 3-isopropyloxypropylene oxide, 3-isopropoxy-1,2-epoxypropane, isopropyl epoxypropyl ether, propane, 1,2-epoxy-3-isopropoxy, oxirane, 1-methylethoxy methyl
PubChem CID 19920
IUPAC Name 2-(propan-2-yloxymethyl)oxirane
SMILES CC(C)OCC1CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
G025525ML
View Documents
TCI America
G025525ML
25 mL
Each for $106.25
Only null left
 
G0255500ML
View Documents
TCI America
G0255500ML
500 mL
Each for $296.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4016-14-2
116.16
NWLUZGJDEZBBRH-UHFFFAOYSA-N
19920
CC(C)OCC1CO1
C6H12O2
MFCD00005139
glycidyl isopropyl ether, isopropyl glycidyl ether, 2,3-epoxypropyl isopropyl ether, isopropoxymethyl oxirane, 2-isopropoxymethyl oxirane, 3-isopropyloxypropylene oxide, 3-isopropoxy-1,2-epoxypropane, isopropyl epoxypropyl ether, propane, 1,2-epoxy-3-isopropoxy, oxirane, 1-methylethoxy methyl
2-(propan-2-yloxymethyl)oxirane

Specifications

4016-14-2
132°C
MFCD00005139
3271
NWLUZGJDEZBBRH-UHFFFAOYSA-N
2-(propan-2-yloxymethyl)oxirane
19920
≥96.0% (GC)
Glycidyl Isopropyl Ether
Colorless
C6H12O2
25 mL
glycidyl isopropyl ether, isopropyl glycidyl ether, 2,3-epoxypropyl isopropyl ether, isopropoxymethyl oxirane, 2-isopropoxymethyl oxirane, 3-isopropyloxypropylene oxide, 3-isopropoxy-1,2-epoxypropane, isopropyl epoxypropyl ether, propane, 1,2-epoxy-3-isopropoxy, oxirane, 1-methylethoxy methyl
CC(C)OCC1CO1
116.16
116.16
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TZ3500000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'