missing translation for 'onlineSavingsMsg'
Learn More

beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%

Catalog No. AC410991000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410991000 100 g Glass bottle
AC410990250 25 g Glass bottle
AC410995000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC410991000 Supplier Thermo Scientific Chemicals Supplier No. 410991000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%, C3H7Na2O6P.5H2O, CAS Number-13408-09-8 | CAS: 13408-09-8 | C3H19Na2O11P | 308.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13408-09-8
Molecular Formula C3H19Na2O11P
Molecular Weight (g/mol) 308.13
MDL Number MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135
InChI Key VWVLGHQKEJESPW-UHFFFAOYSA-N
Synonym unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate
PubChem CID 131846916
IUPAC Name 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate
SMILES O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 102.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH2OHCH(OPO3Na2)CH2OH·5H2O
Quantity 100 g
Solubility Information Solubility in water: freely soluble.
Formula Weight 306.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material β-Glycerophosphoric acid, disodium salt pentahydrate
Show More Show Less
Hazard Category

RUO – Research Use Only