missing translation for 'onlineSavingsMsg'
Learn More

Glycerol tributyrate, 97%

Catalog No. AAA1183018
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1183018 50 g
AAA1183030 250 g
AAA118300B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AAA1183018 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' A1183018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Glycerol tributyrate, 98%, C15H26O6, CAS Number-60-01-5, tri-n-butyrin, glyceryl tributyrate, propane-1,2,3-triyl tributyrate, glycerol tributyrate, glycerol tributanoate, glyceroltributyrin, tributyrin, butyrin, butyryl triglyceride, tributin, 1000g, 1714746, 307 deg.C to 308 deg.C, CHEBI:35020 | CAS: 60-01-5 | C15H26O6 | 302.367 g/mol

Glycerol tributyrate is a triglyceride that may inhibit cell growth and induce cell differentiation. It is used to to induce differentiation of malignant cells followed by arrest of cell growth and apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycerol tributyrate is a triglyceride that may inhibit cell growth and induce cell differentiation. It is used to to induce differentiation of malignant cells followed by arrest of cell growth and apoptosis.

Solubility
Not miscible in water.

Notes
Incompatible with strong oxidizing agents, Strong acids, Strong bases. Use with adequate ventilation.

Chemical Identifiers

CAS 60-01-5
Molecular Formula C15H26O6
Molecular Weight (g/mol) 302.367
MDL Number MFCD00009392
InChI Key UYXTWWCETRIEDR-UHFFFAOYSA-N
Synonym tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin
PubChem CID 6050
ChEBI CHEBI:35020
IUPAC Name 2,3-di(butanoyloxy)propyl butanoate
SMILES CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC

Specifications

Melting Point -75°C
Density 1.032
Boiling Point 307°C to 308°C
Flash Point 174°C (345°F)
Refractive Index 1.4358
Quantity 50 g
Beilstein 1714746
Merck Index 14,9620
Solubility Information Not miscible in water.
Formula Weight 302.37
Percent Purity 97%
Chemical Name or Material Glycerol tributyrate
Show More Show Less
EINECSNumber 200-451-5
RTECSNumber ET7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only