missing translation for 'onlineSavingsMsg'
Learn More

Glycerol triacetate, 99%

CAS: 102-76-1 | C9H14O6 | 218.205 g/mol

Supplier:  Thermo Scientific Chemicals A171320E

Quantity 2500 g

 View more versions of this product

Catalog No. AAA171320E


Only null left

Description

Description

Glycerol triacetate is used as a food additive and as a solvent in flavorings. It acts as an excipient in pharmaceutical products where it is used as a humectant and a plasticizer. It is also used as a fuel additive, as an antiknock agent, adhesives and sealant chemicals, fillers, intermediates and process regulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycerol triacetate is used as a food additive and as a solvent in flavorings. It acts as an excipient in pharmaceutical products where it is used as a humectant and a plasticizer. It is also used as a fuel additive, as an antiknock agent, adhesives and sealant chemicals, fillers, intermediates and process regulators.

Solubility
Slightly miscible with water, carbon tetrachloride and carbondisulfide. Miscible with acetone, ethanol, benzene and chloroform.

Notes
Heat sensitive. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

102-76-1
218.205
URAYPUMNDPQOKB-UHFFFAOYSA-N
5541
2,3-diacetyloxypropyl acetate
C9H14O6
MFCD00008716
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine
CHEBI:9661
CC(=O)OCC(COC(=O)C)OC(=O)C

Specifications

Glycerol triacetate
2°C to 4°C
257°C to 259°C
Petroleum-like
C9H14O6
MFCD00008716
Slightly miscible with water,carbon tetrachloride and carbondisulfide. Miscible with acetone,ethanol,benzene and chloroform.
CC(=O)OCC(COC(=O)C)OC(=O)C
218.205
CHEBI:9661
99%
102-76-1
1.155
149°C (300°F)
99%
2500 g
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine
URAYPUMNDPQOKB-UHFFFAOYSA-N
2,3-diacetyloxypropyl acetate
5541
218.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-051-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions