missing translation for 'onlineSavingsMsg'
Learn More

Glycerol monostearate, purified

Catalog No. AA43883A7
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43883A7 5 kg
AA4388330 250 g
AA43883A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43883A7 Supplier Thermo Scientific Chemicals Supplier No. 043883A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31566-31-1 | C21H42O4 | 358.563 g/mol

Glycerol monostearate is used as an emulsifier, resin lubricant, opacifier, emollient, bodying agent in a variety of cosmetic formulations for skincare and haircare. It is also used as a thickening, anti-caking and preservative agent. It is also useful for preventing ice creams from drying out or being too sweet. It is further used as a foaming agent for the foam-mat drying of papaya. It is also used as an anti-staling agent in bread.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31566-31-1
Molecular Formula C21H42O4
Molecular Weight (g/mol) 358.563
MDL Number MFCD00036186
InChI Key VBICKXHEKHSIBG-UHFFFAOYSA-N
Synonym glyceryl monostearate, monostearin, glyceryl stearate, 1-stearoyl-rac-glycerol, tegin, 1-monostearin, 1-glyceryl stearate, glycerol monostearate, 1-monostearoylglycerol, octadecanoic acid, 2,3-dihydroxypropyl ester
PubChem CID 24699
ChEBI CHEBI:75555
IUPAC Name 2,3-dihydroxypropyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(CO)O

Specifications

Quantity 5 kg
Merck Index 14,4489
Solubility Information Soluble in hot organic solvents;Soluble in hot water; Slightly soluble in ethanol; Insoluble in aliphatic solvents.
Formula Weight 358.56
Physical Form Powder
Chemical Name or Material Glycerol monostearate, Purified
EINECSNumber 250-705-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only