missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol Formal (mixture of 1,3-Dioxan-5-ol and 4-Hydroxymethyldioxolane) 98.0+%, TCI America™
$61.51 - $367.22
Chemical Identifiers
| CAS | 99569-11-6 |
|---|---|
| Molecular Formula | C8H16O6 |
| Molecular Weight (g/mol) | 208.21 |
| MDL Number | MFCD00003218 |
| InChI Key | JIUMSISXCKWZTA-UHFFFAOYSA-N |
| PubChem CID | 251541 |
| IUPAC Name | 1,3-dioxan-5-ol;1,3-dioxolan-4-ylmethanol |
| SMILES | C1C(COCO1)O.C1C(OCO1)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G024425ML
|
TCI America
G024425ML |
25 mL |
Each for $61.51
|
|
|||||
|
G0244500ML
|
TCI America
G0244500ML |
500 mL |
Each for $367.22
|
|
|||||
Chemical Identifiers
| 99569-11-6 | |
| 208.21 | |
| JIUMSISXCKWZTA-UHFFFAOYSA-N | |
| 1,3-dioxan-5-ol;1,3-dioxolan-4-ylmethanol |
| C8H16O6 | |
| MFCD00003218 | |
| 251541 | |
| C1C(COCO1)O.C1C(OCO1)CO |
Specifications
| 99569-11-6 | |
| 97°C | |
| MFCD00003218 | |
| JIUMSISXCKWZTA-UHFFFAOYSA-N | |
| 1,3-dioxan-5-ol;1,3-dioxolan-4-ylmethanol | |
| 251541 | |
| ≥98.0% (GC) | |
| Glycerol Formal (mixture of 1,3-Dioxan-5-ol and 4-Hydroxymethyldioxolane) |
| Yellow | |
| C8H16O6 | |
| 25 mL | |
| C1C(COCO1)O.C1C(OCO1)CO | |
| 208.21 | |
| 104.11 | |
| Liquid |
Safety and Handling
TSCA : No