missing translation for 'onlineSavingsMsg'
Learn More

Glycerol formal, 99+%, mixture of isomers, stabilized

Catalog No. AC259770050
Change view
Click to view available options
Quantity:
5 mL
250 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC259770050 5 mL Glass bottle
AC259772500 250 mL Glass bottle
AC259775000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC259770050 Supplier Thermo Scientific Chemicals Supplier No. 259770050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4740-78-7 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Glycerol formal
Name Note 99+%
CAS 4740-78-7, 5464-28-8
Density 1.2100g/mL
pH 4.0 to 6.5 (10% soln.)
Boiling Point 193.0°C to 195.0°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. Sum of alpha-and beta-isomers (GC)
Packaging Glass bottle
Molecular Formula C4H8O3
Refractive Index 1.4500 to 1.4520
Quantity 5 mL
MDL Number MFCD00003218
Specific Gravity 1.21
Merck Index 14, 4485
Solubility Information (50% in water) Clear
Molecular Weight (g/mol) 104.11
Viscosity 16 mPa.s (20°C)
Formula Weight 104.11
Percent Purity 99+%
Physical Form Liquid
Show More Show Less

RUO – Research Use Only