missing translation for 'onlineSavingsMsg'
Learn More

Glycerol/Ethanol Solution, Reagents

Catalog No. 01335165
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01335165 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335165 Supplier Labchem Inc Supplier No. CG1000004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glycerol/Ethanol Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Glycerol/Ethanol Solution
CAS 64-17-5, 67-63-0, 67-56-1
Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.9545
Synonym 1,2,3-Propanetriol/Ethyl alcohol
Grade Reagent