missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol alpha,alpha'-Diallyl Ether 98.0+%, TCI America™
$159.49 - $1175.25
Chemical Identifiers
| CAS | 17018-07-4 |
|---|---|
| Molecular Formula | C9H16O3 |
| Molecular Weight (g/mol) | 172.224 |
| MDL Number | MFCD00085245 |
| InChI Key | OFLGYLDXUIXHQY-UHFFFAOYSA-N |
| Synonym | 1,3-Diallyloxy-2-propanol, GDAE |
| PubChem CID | 86908 |
| IUPAC Name | 1,3-bis(prop-2-enoxy)propan-2-ol |
| SMILES | C=CCOCC(COCC=C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D214625ML
|
TCI America
D214625ML |
25 mL |
Each for $159.49
|
|
|||||
|
D2146500ML
|
TCI America
D2146500ML |
500 mL |
Each for $1,175.25
|
|
|||||
Chemical Identifiers
| 17018-07-4 | |
| 172.224 | |
| OFLGYLDXUIXHQY-UHFFFAOYSA-N | |
| 86908 | |
| C=CCOCC(COCC=C)O |
| C9H16O3 | |
| MFCD00085245 | |
| 1,3-Diallyloxy-2-propanol, GDAE | |
| 1,3-bis(prop-2-enoxy)propan-2-ol |
Specifications
| 17018-07-4 | |
| 106°C | |
| MFCD00085245 | |
| 1,3-Diallyloxy-2-propanol, GDAE | |
| C=CCOCC(COCC=C)O | |
| 172.224 | |
| 172.22 | |
| Liquid |
| Colorless | |
| C9H16O3 | |
| 25 mL | |
| OFLGYLDXUIXHQY-UHFFFAOYSA-N | |
| 1,3-bis(prop-2-enoxy)propan-2-ol | |
| 86908 | |
| ≥98.0% (GC) | |
| Glycerol alpha,alpha′-Diallyl Ether |
Safety and Handling
EINECSNumber : (2)-0428
RTECSNumber : UA7380500
TSCA : Yes