missing translation for 'onlineSavingsMsg'
Learn More

Glycerol alpha,alpha'-Diallyl Ether 98.0+%, TCI America™

Catalog No. D2146500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2146500ML 500 mL
D214625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2146500ML Supplier TCI America Supplier No. D2146500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17018-07-4
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
MDL Number MFCD00085245
InChI Key OFLGYLDXUIXHQY-UHFFFAOYSA-N
Synonym 1,3-Diallyloxy-2-propanol, GDAE
PubChem CID 86908
IUPAC Name 1,3-bis(prop-2-enoxy)propan-2-ol
SMILES C=CCOCC(COCC=C)O

Specifications

Color Colorless
Boiling Point 106°C
Quantity 500 mL
Formula Weight 172.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycerol alpha,alpha′-Diallyl Ether
EINECSNumber (2)-0428
RTECSNumber UA7380500
TSCA Yes