missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Gly-Phe beta-naphthylamide
Description
Gly-Phe beta-naphthylamide is used to accumulate inside lysosomes leading to their destruction. Studies report that Gly-Phe-β-naphthylamide significantly inhibits the activation of caspase-8, and has also been used to study recombinant cathepsin C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Purification Method | HPLC |
| Common Name | Glycylphenylalanine 2-naphthylamide |
| Content And Storage | -30°C to -10°C |
| Format | Crystalline |
| For Use With (Application) | Substrate for cathepsin C |
| Molecular Weight (g/mol) | 347.41 |
| Product Type | Gly-Phe beta-naphthylamide |
| Quantity | 100 mg |
| Regulatory Status | RUO - research use only |
| Sequence | (S)-2-(2-Aminoacetamido)-N-(naphthalen-2-yl)-3-phenylpropanamide |
RUO – Research Use Only