missing translation for 'onlineSavingsMsg'
Learn More

Glutathione, Reduced, Immobilized on Agarose CL-4B, For affinity chromatography, Powder (lyophilized), MilliporeSigma™ Supelco™

the lactose stabilizer must be removed prior to use by washing the gel onto filter with water or buffer; the reduced glutathione is attached through the sulfur to epoxy-activated 4% cross-linked beaded agarose (C10-spacer)

$2539.31 - $2539.31

Specifications

Grade For affinity chromatography
Physical Form Lyophilized Powder
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111003043
View Documents
Merck Emd Millipore
4973950ML
50 mL
Each for $2,539.31
Only null left
 
Description

Description

Specifications

Specifications

For affinity chromatography
Lyophilized Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'