missing translation for 'onlineSavingsMsg'
Learn More

Glutathione reduced form 97.0+%, TCI America™

Catalog No. p-7130133
Change view
Click to view available options
Quantity:
1 g
10 g
Catalog No. Quantity
G00741G 1 g
G007410G 10 g
2 options

Catalog No. G00741G

Supplier: TCI America G00741G

Only null left

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Melting Point 195°C
Color White
Quantity 1 g
Formula Weight 307.32
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glutathione reduced form
EINECSNumber (9)-1571
RTECSNumber MC0556000
TSCA Yes