missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glutathione Reduced Crystalline MP Biomedicals
Manufacturer: Mp Biomedicals 0210181450
Specifications
Glutathione | |
50g | |
70-18-8 | |
C10H17N3O6S | |
deltathione, glutathion, glutathione, glutathione-sh, glutinal, isethion, l-glutathione, neuthion, reduced glutathione, tathion | |
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N | |
307.321 | |
CHEBI:16856 | |
Powder |
White | |
Reduced | |
>98% | |
MFCD00065939 | |
RWSXRVCMGQZWBV-WDSKDSINSA-N | |
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid | |
124886 | |
307.3 |
Chemical Identifiers
70-18-8 | |
307.321 | |
RWSXRVCMGQZWBV-WDSKDSINSA-N | |
124886 | |
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid |
C10H17N3O6S | |
MFCD00065939 | |
deltathione, glutathion, glutathione, glutathione-sh, glutinal, isethion, l-glutathione, neuthion, reduced glutathione, tathion | |
CHEBI:16856 | |
C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N |
Safety and Handling
EINECSNumber : 200-725-4
Recommended Storage : Store at 4°C. Store under nitrogen.