missing translation for 'onlineSavingsMsg'
Learn More

Glutathione Reduced Crystalline MP Biomedicals

Catalog No. ICN10181425
Change view
Click to view available options
Quantity:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10181425 25 g
ICN10181450 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10181425 Supplier MP Biomedicals Inc Supplier No. 0210181425

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.321
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Color White
Assay Percent Range >98%
Quantity 25 g
Formula Weight 307.3
Physical Form Powder
Chemical Name or Material Glutathione, Reduced
EINECSNumber 200-725-4
Recommended Storage Store at 4°C. Store under nitrogen.