missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione oxidized form 98.0+%, TCI America™
Supplier: TCI America G00731G
Specifications
| Glutathione oxidized form | |
| White-Yellow | |
| MFCD00150701 | |
| oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized | |
| N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O | |
| 612.63 | |
| CHEBI:17858 | |
| ≥98.0% (HPLC) |
| 27025-41-8 | |
| C20H32N6O12S2 | |
| 1 g | |
| YPZRWBKMTBYPTK-IOKZUGQQNA-N | |
| (2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid | |
| 65359 | |
| 612.63 | |
| Crystalline Powder |
Chemical Identifiers
| 27025-41-8 | |
| 612.63 | |
| YPZRWBKMTBYPTK-IOKZUGQQNA-N | |
| 65359 | |
| (2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid |
| C20H32N6O12S2 | |
| MFCD00150701 | |
| oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized | |
| CHEBI:17858 | |
| N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O |
Safety and Handling
EINECSNumber : (9)-1571
RTECSNumber : MC0556500
TSCA : No