missing translation for 'onlineSavingsMsg'
Learn More

Glutaryl Chloride 95.0+%, TCI America™

Catalog No. G020225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G020225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G020225ML Supplier TCI America Supplier No. G020225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2873-74-7
Molecular Formula C5H6Cl2O2
Molecular Weight (g/mol) 169.001
MDL Number MFCD00000755
InChI Key YVOFTMXWTWHRBH-UHFFFAOYSA-N
Synonym glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride
PubChem CID 17887
IUPAC Name pentanedioyl dichloride
SMILES C(CC(=O)Cl)CC(=O)Cl

Specifications

Color Yellow
Boiling Point 218°C
Quantity 25 mL
UN Number 2922
Formula Weight 169.00
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Glutaryl Chloride
RTECSNumber SA2583000
TSCA Yes