missing translation for 'onlineSavingsMsg'
Learn More

Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified

Catalog No. p-3778704
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233280250 25 mL Glass bottle
AC233281000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC233280250 Supplier Thermo Scientific Chemicals Supplier No. 233280250
Only null left

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Glutaric dialdehyde, electron microscopy grade
Name Note Purified
CAS 111-30-8, 7732-18-5
Melting Point -5.0°C
Density 1.0600g/mL
pH 3.1 to 4.5 (25°C)
Boiling Point 101°C
Packaging Glass bottle
Molecular Formula C5H8O2
Refractive Index 1.373
Linear Formula OHC(CH2)3CHO
Quantity 25 mL
MDL Number MFCD00007025
Beilstein 01, 776
Fieser 01,411
Specific Gravity 1.06
Merck Index 15, 4508
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
Solubility Information Solubility in water: soluble
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILES O=CCCCC=O
IUPAC Name pentanedial
Molecular Weight (g/mol) 100.12
PubChem CID 3485
ChEBI CHEBI:64276
Formula Weight 100.12
Percent Purity 25 to 27 wt%
Grade E.M.
Physical Form Solution
Show More Show Less

RUO – Research Use Only