missing translation for 'onlineSavingsMsg'
Learn More

Glutaric Anhydride 98.0+%, TCI America™

Catalog No. G0071500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0071500G 500 g
G007125G 25 g
G0071100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G0071500G Supplier TCI America Supplier No. G0071500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-55-4
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00006679
InChI Key VANNPISTIUFMLH-UHFFFAOYSA-N
Synonym glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
PubChem CID 7940
IUPAC Name oxane-2,6-dione
SMILES C1CC(=O)OC(=O)C1

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 146°C
Quantity 500 g
Formula Weight 114.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glutaric Anhydride
EINECSNumber (2)-3431
RTECSNumber MA3850000
TSCA Yes