missing translation for 'onlineSavingsMsg'
Learn More

StatLab Glutaraldehyde Buffer 2% pH 7.3

Catalog No. 22000020
Change view
Click to view available options
Quantity:
100 mL
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22000020 100 mL
22000021 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22000020 Supplier StatLab Supplier No. BUG0526
Harmful if swallowed, in contact with skin, or if inhaled

Glutaraldehyde Buffer 2% pH 7.3

  • Glutaraldehyde Buffer 2% pH 7.3
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
CAS 111-30-8, 124-65-2
Color Clear Colorless
pH 7.3
Quantity 100 mL
Grade Laboratory